Search
Search

Galnt1| Gene Symbol: Galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160404.1 | NP_001153876.1 | 4 | 445 | ||
| NM_013814.3 | NP_038842.3 | 4 | 533 | |||
| GenBank mRNA | AK083391.1 | - | 4 | 446 | ||
| BC056215.1 | - | 4 | 450 | 30019829 | ||
| BC090962.1 | - | 3 | 323 | 30626890 | ||
| BY711053.1 | - | 4 | 495 | |||
| U73820.1 | - | 2 | 198 | |||