Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARF1 SIRNA Assays ›
          Gene Symbol:
          Arf1
          Gene Name:
          ADP-ribosylation factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 59211411 - 59228267 on Build GRCm38
          UniGene ID:
          Mm.371546
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159706

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Arf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130408.1 NP_001123880.1 5 1956
          NM_007476.3 NP_031502.1 5 1764
          GenBank mRNA AK077542.1 - 5 1712
          AK148458.1 - 5 1728
          AK148873.1 - 5 1718
          AK150092.1 - 5 1737
          AK150699.1 - 5 1715
          AK151223.1 - 5 1715
          AK151225.1 - 5 1738
          AK151696.1 - 5 1690
          AK151821.1 - 5 1712
          AK152382.1 - 5 1712
          AK166628.1 - 4 1579
          AK167000.1 - 5 1693
          AK167129.1 - 5 1727
          AK167543.1 - 5 1714
          AK167931.1 - 5 1727
          AK168686.1 - 5 1714
          AK169166.1 - 5 1712
          AK170586.1 - 5 1689
          BC021403.1 - 4 1707
          BC031986.1 - 4 1681 4012689

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor internalization
          transport
          cellular copper ion homeostasis
          actin filament organization
          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          regulation of Arp2/3 complex-mediated actin nucleation
          very-low-density lipoprotein particle assembly
          positive regulation of catalytic activity
          positive regulation of endocytosis
          positive regulation of calcium ion-dependent exocytosis
          positive regulation of protein secretion
          Golgi to transport vesicle transport
          long term synaptic depression
          positive regulation of dendritic spine development
          synaptic vesicle budding
          dendritic spine organization
          lysosomal membrane organization
          positive regulation of sodium ion transmembrane transport
          positive regulation of late endosome to lysosome transport
          positive regulation of ER to Golgi vesicle-mediated transport
          regulation of phospholipid metabolic process
          nucleotide binding
          magnesium ion binding
          protein binding
          GTP binding
          GDP binding
          protein homodimerization activity
          poly(A) RNA binding
          phospholipase D activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0