Search
Search

Bmi1| Gene Symbol: Bmi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007552.4 | NP_031578.2 | 8 | 958 | ||
| GenBank mRNA | BC053708.1 | - | 8 | 584 | 30074540 | |
| BC056384.1 | - | 7 | 488 | 6854104 | ||
| DQ337175.1 | - | 7 | 487 | |||
| M64279.1 | - | 8 | 958 | |||