Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAV3 SIRNA Assays ›
Gene Symbol:
Cav3
Gene Name:
caveolin 3
Gene Aliases:
AI385751, Cav-3, M-cav
Chromosome Location:
Chr.6: 112459505 - 112472872 on Build GRCm38
UniGene ID:
Mm.3924
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
160017
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cav3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007617.3
NP_031643.1
2
473
GenBank mRNA
BC024383.1
-
2
483
4988598
CJ057698.1
-
2
250
U36579.1
-
2
287
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane repair
regulation of heart rate
negative regulation of protein kinase activity
triglyceride metabolic process
endocytosis
plasma membrane organization
actin filament organization
positive regulation of cytosolic calcium ion concentration
myoblast fusion
regulation of heart contraction
protein localization
positive regulation of cell proliferation
negative regulation of cardiac muscle hypertrophy
positive regulation of myotube differentiation
regulation of skeletal muscle contraction
myotube differentiation
regulation of transforming growth factor beta receptor signaling pathway
positive regulation of microtubule polymerization
cytoplasmic microtubule organization
membrane raft organization
detection of muscle stretch
regulation of signal transduction by receptor internalization
regulation of membrane potential
glucose homeostasis
cholesterol homeostasis
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
negative regulation of cell size
muscle cell cellular homeostasis
negative regulation of nitric-oxide synthase activity
regulation of nerve growth factor receptor activity
nucleus localization
regulation of protein kinase B signaling
regulation of calcium ion transport
negative regulation of calcium ion transport
cardiac muscle cell development
regulation of cardiac muscle contraction
negative regulation of sarcomere organization
regulation of ventricular cardiac muscle cell membrane repolarization
heart trabecula formation
regulation of ventricular cardiac muscle cell membrane depolarization
regulation of branching involved in mammary gland duct morphogenesis
negative regulation of cell growth involved in cardiac muscle cell development
caveola assembly
protein localization to plasma membrane
ventricular cardiac muscle cell action potential
establishment of protein localization to plasma membrane
regulation of calcium ion import
regulation of p38MAPK cascade
regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
calcium channel regulator activity
protein binding
protein C-terminus binding
sodium channel regulator activity
potassium channel inhibitor activity
enzyme binding
protein complex binding
protein complex scaffold
alpha-tubulin binding
ion channel binding
nitric-oxide synthase binding
connexin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0