Search
Search

Cbx2| Gene Symbol: Cbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007623.2 | NP_031649.2 | 5 | 380 | ||
| GenBank mRNA | AK157869.1 | - | 5 | 356 | ||
| BC035199.1 | - | 5 | 380 | 5137505 | ||
| X62537.1 | - | 5 | 447 | |||
| Y13274.1 | - | 5 | 304 | |||