Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK5 SIRNA Assays ›
Gene Symbol:
Cdk5
Gene Name:
cyclin-dependent kinase 5
Gene Aliases:
AW048668, Crk6
Chromosome Location:
Chr.5: 24407597 - 24423591 on Build GRCm38
UniGene ID:
Mm.298798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
160138
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cdk5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007668.3
NP_031694.1
12
1207
GenBank mRNA
AK051790.1
-
12
1207
BC052007.1
-
12
1181
5706932
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
synaptic transmission, dopaminergic
protein phosphorylation
intracellular protein transport
exocytosis
nucleocytoplasmic transport
apoptotic process
cell cycle
cell-matrix adhesion
nervous system development
axonogenesis
synapse assembly
skeletal muscle tissue development
motor neuron axon guidance
associative learning
visual learning
response to wounding
Schwann cell development
phosphorylation
cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
telencephalon development
cerebellum development
cerebellar cortex development
cerebellar cortex formation
hippocampus development
layer formation in cerebral cortex
central nervous system neuron development
cerebral cortex development
corpus callosum development
neuron differentiation
regulation of cell migration
negative regulation of axon extension
cortical actin cytoskeleton organization
forebrain development
neuron projection development
negative regulation of protein ubiquitination
negative regulation of synaptic plasticity
positive regulation of protein binding
receptor catabolic process
serine phosphorylation of STAT3 protein
synaptic transmission, glutamatergic
protein localization to synapse
response to cocaine
receptor clustering
positive regulation of neuron apoptotic process
regulated exocytosis
negative regulation of cell cycle
positive regulation of protein kinase activity
negative regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of calcium ion-dependent exocytosis
protein autophosphorylation
negative regulation of protein export from nucleus
behavioral response to cocaine
regulation of synaptic plasticity
synaptic vesicle endocytosis
rhythmic process
oligodendrocyte differentiation
neuron projection morphogenesis
dendrite morphogenesis
cell division
neuron apoptotic process
regulation of cell cycle
regulation of postsynaptic membrane potential
excitatory postsynaptic potential
regulation of dendritic spine morphogenesis
calcium ion import
positive regulation of protein targeting to membrane
negative regulation of neuron death
positive regulation of receptor activity
nucleotide binding
p53 binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ErbB-2 class receptor binding
protein binding
ATP binding
cytoskeletal protein binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
acetylcholine receptor activator activity
ErbB-3 class receptor binding
ephrin receptor binding
tau-protein kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0