Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM2 SIRNA Assays ›
          Gene Symbol:
          Dnm2
          Gene Name:
          dynamin 2
          Gene Aliases:
          Dyn2, Udnm, b2b2159Clo
          Chromosome Location:
          Chr.9: 21424908 - 21507759 on Build GRCm38
          UniGene ID:
          Mm.433257
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 160566

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dnm2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039520.2 NP_001034609.1 3, 4 575
          NM_001253893.1 NP_001240822.1 3, 4 575
          NM_001253894.1 NP_001240823.1 3, 4 575
          NM_007871.2 NP_031897.2 3, 4 575
          GenBank mRNA AK005012.2 - 3, 4 551
          AK015410.1 - 2, 3 240
          AK150046.1 - 3, 4 538
          AK154375.1 - 2, 3 239
          AK157844.1 - 3, 4 545
          AK170574.1 - 3, 4 551
          AK171049.1 - 3, 4 529
          AK171566.1 - 3, 4 539
          AK172231.1 - 3, 4 556
          L31398.1 - 3, 4 515

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013199, XM_006242614, XM_006242611, XM_006242613, XM_008765917, and others.

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor internalization
          Golgi to plasma membrane transport
          endocytosis
          receptor-mediated endocytosis
          positive regulation of lamellipodium assembly
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of axon extension
          receptor internalization
          transferrin transport
          regulation of Rac protein signal transduction
          macropinocytosis
          positive regulation of nitric oxide biosynthetic process
          positive regulation of endocytosis
          neuron projection morphogenesis
          positive regulation of phagocytosis
          cilium morphogenesis
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to dopamine
          regulation of Golgi organization
          positive regulation of sodium:potassium-exchanging ATPase activity
          negative regulation of membrane tubulation
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          hydrolase activity
          SH3 domain binding
          protein kinase binding
          D2 dopamine receptor binding
          protein complex binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          WW domain binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0