Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALOX12B SIRNA Assays ›
Gene Symbol:
Alox12b
Gene Name:
arachidonate 12-lipoxygenase, 12R type
Gene Aliases:
Aloxe2, e-LOX2
Chromosome Location:
Chr.11: 69157072 - 69169791 on Build GRCm38
UniGene ID:
Mm.340329
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
160640
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Alox12b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009659.2
NP_033789.1
1
281
GenBank mRNA
AF059251.1
-
1
159
BC113149.1
-
1
245
40054352
Y14334.1
-
1
281
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001039377, AY903232, AY903233, AY903231.
Panther Classification:
Molecular Function -
oxygenase
Gene Ontology Categories:
Function(s)
Process(es)
protein lipidation
lipid metabolic process
fatty acid metabolic process
sphingolipid metabolic process
positive regulation of gene expression
arachidonic acid metabolic process
lipoxygenase pathway
positive regulation of MAPK cascade
linoleic acid metabolic process
ceramide biosynthetic process
hepoxilin biosynthetic process
oxidation-reduction process
establishment of skin barrier
positive regulation of mucus secretion
bicellular tight junction assembly
arachidonate 12-lipoxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
2,3-dihydroxy DDT 1,2-dioxygenase activity
phenanthrene dioxygenase activity
2,2',3-trihydroxybiphenyl dioxygenase activity
1,2-dihydroxyfluorene 1,1-alpha-dioxygenase activity
5,6-dihydroxy-3-methyl-2-oxo-1,2-dihydroquinoline dioxygenase activity
1,1-dichloro-2-(dihydroxy-4-chlorophenyl)-(4-chlorophenyl)ethene 1,2-dioxygenase activity
protocatechuate 3,4-dioxygenase type II activity
2'-aminobiphenyl-2,3-diol 1,2-dioxygenase activity
3,4-dihydroxyfluorene 4,4-alpha-dioxygenase activity
2,3-dihydroxy-ethylbenzene 1,2-dioxygenase activity
carbazole 1,9a-dioxygenase activity
dihydroxydibenzothiophene dioxygenase activity
1,2-dihydroxynaphthalene-6-sulfonate 1,8a-dioxygenase activity
styrene dioxygenase activity
3,4-dihydroxyphenanthrene dioxygenase activity
hydroquinone 1,2-dioxygenase activity
p-cumate 2,3-dioxygenase activity
2,3-dihydroxy-p-cumate dioxygenase activity
3,5-dichlorocatechol 1,2-dioxygenase activity
2-aminophenol 1,6-dioxygenase activity
2,6-dichloro-p-hydroquinone 1,2-dioxygenase activity
chlorocatechol 1,2-dioxygenase activity
catechol dioxygenase activity
dihydroxyfluorene dioxygenase activity
5-aminosalicylate dioxygenase activity
3-hydroxy-2-naphthoate 2,3-dioxygenase activity
benzo(a)pyrene 11,12-dioxygenase activity
benzo(a)pyrene 4,5-dioxygenase activity
4,5-dihydroxybenzo(a)pyrene dioxygenase activity
benzo(a)pyrene 9,10-dioxygenase activity
9,10-dihydroxybenzo(a)pyrene dioxygenase activity
benzo(a)pyrene 7,8-dioxygenase activity
7,8-dihydroxy benzo(a)pyrene dioxygenase activity
1,2-dihydroxy-5,6,7,8-tetrahydronaphthalene extradiol dioxygenase activity
2-mercaptobenzothiazole dioxygenase activity
pyridine-3,4-diol dioxygenase activity
pyrene dioxygenase activity
4,5-dihydroxypyrene dioxygenase activity
phenanthrene-4-carboxylate dioxygenase activity
tetrachlorobenzene dioxygenase activity
4,6-dichloro-3-methylcatechol 1,2-dioxygenase activity
2,3-dihydroxydiphenyl ether dioxygenase activity
diphenyl ether 1,2-dioxygenase activity
arachidonate 8(S)-lipoxygenase activity
metal ion binding
4-hydroxycatechol 1,2-dioxygenase activity
dioxygenase activity
linoleate 9S-lipoxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0