Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALOX12B SIRNA Assays ›
          Gene Symbol:
          Alox12b
          Gene Name:
          arachidonate 12-lipoxygenase, 12R type
          Gene Aliases:
          Aloxe2, e-LOX2
          Chromosome Location:
          Chr.11: 69157072 - 69169791 on Build GRCm38
          UniGene ID:
          Mm.340329
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 160641

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Alox12b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009659.2 NP_033789.1 3, 4 606
          GenBank mRNA AF059251.1 - 3, 4 484
          BC113149.1 - 3, 4 570 40054352
          Y14334.1 - 3, 4 606

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein lipidation
          lipid metabolic process
          fatty acid metabolic process
          sphingolipid metabolic process
          positive regulation of gene expression
          arachidonic acid metabolic process
          lipoxygenase pathway
          positive regulation of MAPK cascade
          linoleic acid metabolic process
          ceramide biosynthetic process
          hepoxilin biosynthetic process
          oxidation-reduction process
          establishment of skin barrier
          positive regulation of mucus secretion
          bicellular tight junction assembly
          arachidonate 12-lipoxygenase activity
          iron ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          2,3-dihydroxy DDT 1,2-dioxygenase activity
          phenanthrene dioxygenase activity
          2,2',3-trihydroxybiphenyl dioxygenase activity
          1,2-dihydroxyfluorene 1,1-alpha-dioxygenase activity
          5,6-dihydroxy-3-methyl-2-oxo-1,2-dihydroquinoline dioxygenase activity
          1,1-dichloro-2-(dihydroxy-4-chlorophenyl)-(4-chlorophenyl)ethene 1,2-dioxygenase activity
          protocatechuate 3,4-dioxygenase type II activity
          2'-aminobiphenyl-2,3-diol 1,2-dioxygenase activity
          3,4-dihydroxyfluorene 4,4-alpha-dioxygenase activity
          2,3-dihydroxy-ethylbenzene 1,2-dioxygenase activity
          carbazole 1,9a-dioxygenase activity
          dihydroxydibenzothiophene dioxygenase activity
          1,2-dihydroxynaphthalene-6-sulfonate 1,8a-dioxygenase activity
          styrene dioxygenase activity
          3,4-dihydroxyphenanthrene dioxygenase activity
          hydroquinone 1,2-dioxygenase activity
          p-cumate 2,3-dioxygenase activity
          2,3-dihydroxy-p-cumate dioxygenase activity
          3,5-dichlorocatechol 1,2-dioxygenase activity
          2-aminophenol 1,6-dioxygenase activity
          2,6-dichloro-p-hydroquinone 1,2-dioxygenase activity
          chlorocatechol 1,2-dioxygenase activity
          catechol dioxygenase activity
          dihydroxyfluorene dioxygenase activity
          5-aminosalicylate dioxygenase activity
          3-hydroxy-2-naphthoate 2,3-dioxygenase activity
          benzo(a)pyrene 11,12-dioxygenase activity
          benzo(a)pyrene 4,5-dioxygenase activity
          4,5-dihydroxybenzo(a)pyrene dioxygenase activity
          benzo(a)pyrene 9,10-dioxygenase activity
          9,10-dihydroxybenzo(a)pyrene dioxygenase activity
          benzo(a)pyrene 7,8-dioxygenase activity
          7,8-dihydroxy benzo(a)pyrene dioxygenase activity
          1,2-dihydroxy-5,6,7,8-tetrahydronaphthalene extradiol dioxygenase activity
          2-mercaptobenzothiazole dioxygenase activity
          pyridine-3,4-diol dioxygenase activity
          pyrene dioxygenase activity
          4,5-dihydroxypyrene dioxygenase activity
          phenanthrene-4-carboxylate dioxygenase activity
          tetrachlorobenzene dioxygenase activity
          4,6-dichloro-3-methylcatechol 1,2-dioxygenase activity
          2,3-dihydroxydiphenyl ether dioxygenase activity
          diphenyl ether 1,2-dioxygenase activity
          arachidonate 8(S)-lipoxygenase activity
          metal ion binding
          4-hydroxycatechol 1,2-dioxygenase activity
          dioxygenase activity
          linoleate 9S-lipoxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0