Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1C SIRNA Assays ›
          Gene Symbol:
          Cacna1c
          Gene Name:
          calcium channel, voltage-dependent, L type, alpha 1C subunit
          Gene Aliases:
          Cav1.2, Cchl1a1, D930026N18Rik, MBC, MELC-CC
          Chromosome Location:
          Chr.6: 118587240 - 119198921 on Build GRCm38
          UniGene ID:
          Mm.41628
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 160932

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cacna1c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159533.2 NP_001153005.1 25, 26 3738
          NM_001159534.2 NP_001153006.1 25, 26 3738
          NM_001159535.2 NP_001153007.1 25, 26 3738
          NM_001255997.2 NP_001242926.1 25, 26 3486
          NM_001255998.2 NP_001242927.1 26, 27 3561
          NM_001255999.2 NP_001242928.1 25, 26 3486
          NM_001256000.2 NP_001242929.1 26, 27 3813
          NM_001256001.2 NP_001242930.1 26, 27 3822
          NM_001256002.2 NP_001242931.1 26, 27 3813
          NM_001290335.1 NP_001277264.1 21, 22 2835
          NM_009781.4 NP_033911.2 25, 26 3486
          GenBank mRNA AB259049.1 - 25, 26 3308
          AY728090.1 - 25, 26 3273
          BC138031.1 - 25, 26 3432 8861050
          BC145103.1 - 25, 26 3432
          BC145104.1 - 25, 26 3432 9053652
          BC145105.1 - 25, 26 3432 9053653
          FM872409.1 - 25, 26 3308
          FM872410.1 - 26, 27 3383
          FM872411.1 - 25, 26 3308
          FM872412.1 - 26, 27 3293
          FM872413.1 - 26, 27 3302
          FM872414.1 - 26, 27 3293
          U17869.1 - 21, 22 2835

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          smooth muscle contraction
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          adult walking behavior
          regulation of blood pressure
          visual learning
          calcium ion regulated exocytosis
          regulation of vasoconstriction
          insulin secretion
          growth hormone secretion
          regulation of ion transmembrane transport
          glucose homeostasis
          regulation of organ growth
          transmembrane transport
          smooth muscle contraction involved in micturition
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          high voltage-gated calcium channel activity
          enzyme binding
          protein domain specific binding
          ion channel binding
          metal ion binding
          alpha-actinin binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0