Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK5R1 SIRNA Assays ›
          Gene Symbol:
          Cdk5r1
          Gene Name:
          cyclin-dependent kinase 5, regulatory subunit 1 (p35)
          Gene Aliases:
          Cdk5r, D11Bwg0379e, p25, p35
          Chromosome Location:
          Chr.11: 80477046 - 80481179 on Build GRCm38
          UniGene ID:
          Mm.142275
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 161149

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cdk5r1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009871.2 NP_034001.1 1 2021
          GenBank mRNA AK039999.1 - 2 1812
          AK043203.1 - 1 1997
          AK048617.1 - 1 2000
          AK079557.1 - 1 2024
          AK084003.1 - 1 1979
          AK086621.1 - 1 2050
          AK141549.1 - 1 2034
          AK142868.1 - 1 2047
          AK169667.1 - 1 2025
          BC046823.1 - 1 2006 6400348
          BC050768.1 - 1 2021 6533538
          BC058697.1 - 1 2013 6825809

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          protein phosphorylation
          neuron cell-cell adhesion
          G-protein coupled acetylcholine receptor signaling pathway
          axon guidance
          axonal fasciculation
          brain development
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cerebellum development
          superior olivary nucleus maturation
          hippocampus development
          cerebral cortex radially oriented cell migration
          layer formation in cerebral cortex
          neuron differentiation
          negative regulation of axon extension
          neuron projection development
          regulation of actin cytoskeleton organization
          serine phosphorylation of STAT3 protein
          ionotropic glutamate receptor signaling pathway
          positive regulation of neuron apoptotic process
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          ephrin receptor signaling pathway
          rhythmic process
          regulation of cell cycle
          regulation of dendritic spine morphogenesis
          regulation of microtubule cytoskeleton organization
          positive regulation of cell cycle arrest
          positive regulation of protein serine/threonine kinase activity
          positive regulation of protein targeting to membrane
          cyclin-dependent protein serine/threonine kinase activity
          calcium ion binding
          protein binding
          cytoskeletal protein binding
          kinase activity
          cyclin-dependent protein kinase 5 activator activity
          protein kinase binding
          protein serine/threonine kinase activator activity
          cadherin binding
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0