Search
Search

Dsc1| Gene Symbol: Dsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291804.1 | NP_001278733.1 | 2, 3 | 361 | ||
| NM_013504.5 | NP_038532.3 | 2, 3 | 361 | |||
| GenBank mRNA | AK036893.1 | - | 2, 3 | 247 | ||
| AK079441.1 | - | 2, 3 | 362 | |||
| AK081826.1 | - | 2, 3 | 269 | |||
| BC109161.1 | - | 2, 3 | 198 | 40047883 | ||
| BC109162.1 | - | 2, 3 | 198 | |||
| X97986.1 | - | 2, 3 | 295 | |||