Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP5B SIRNA Assays ›
          Gene Symbol:
          Atp5b
          Gene Name:
          ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 128083307 - 128090388 on Build GRCm38
          UniGene ID:
          Mm.238973
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 161783

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp5b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016774.3 NP_058054.2 5 754
          GenBank mRNA AF030559.1 - 5 753
          AK002621.1 - 5 751
          AK010314.1 - 5 754
          AK083276.1 - 5 757
          AK084009.1 - 5 745
          AK145684.1 - 5 755
          AK148891.1 - 5 756
          AK150599.1 - 5 756
          AK151081.1 - 5 756
          AK151600.1 - 5 753
          AK152730.1 - 5 759
          AK152788.1 - 5 756
          AK152976.1 - 5 756
          AK153099.1 - 5 745
          AK159444.1 - 5 756
          AK159737.1 - 5 759
          AK159978.1 - 5 756
          AK160199.1 - 5 748
          AK160608.1 - 5 756
          AK164383.1 - 5 744
          AK166525.1 - 5 757
          AK166603.1 - 5 756
          AK166979.1 - 5 755
          AK167119.1 - 5 756
          AK167160.1 - 5 746
          AK167728.1 - 5 753
          AK167764.1 - 5 756
          AK168692.1 - 5 746
          AK168941.1 - 5 756
          AK169184.1 - 5 756
          AK178179.1 - 1 96
          AK179339.1 - 3 328
          AK203332.1 - 3 273
          AK205339.1 - 3 279
          BC013253.1 - 5 730
          BC037127.1 - 5 747
          BC046616.1 - 5 737 6494223
          DQ403100.1 - 4 542

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_134364, DQ403101, M57634, FQ230301, FQ216116, and others.

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          osteoblast differentiation
          lipid metabolic process
          ATP biosynthetic process
          transport
          ion transport
          receptor-mediated endocytosis
          negative regulation of cell adhesion involved in substrate-bound cell migration
          ATP synthesis coupled proton transport
          ATP hydrolysis coupled proton transport
          proton transport
          ATP metabolic process
          regulation of intracellular pH
          hydrogen ion transmembrane transport
          nucleotide binding
          calcium ion binding
          ATP binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
          ATPase activity
          lipoprotein particle receptor activity
          MHC class I protein binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0