Search
Search

Fabp4| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024406.2 | NP_077717.1 | 4 | 485 | ||
| GenBank mRNA | AK003143.1 | - | 4 | 486 | ||
| AK088535.1 | - | 4 | 486 | |||
| BC002148.1 | - | 1 | 1900 | 3484864 | ||
| BC054426.1 | - | 4 | 468 | 5029359 | ||
| K02109.1 | - | 4 | 463 | |||
| M28726.1 | - | 4 | 390 | |||