Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACVR2A SIRNA Assays ›
Gene Symbol:
Acvr2a
Gene Name:
activin receptor IIA
Gene Aliases:
ActrIIa, Acvr2, TactrII
Chromosome Location:
Chr.2: 48814109 - 48903264 on Build GRCm38
UniGene ID:
Mm.314338
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
162113
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acvr2a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007396.4
NP_031422.3
11
4201
GenBank mRNA
AK041246.1
-
11
4154
AK141435.1
-
1
1046
AK143186.1
-
12
4300
Back To Top
More Information
Panther Classification:
Molecular Function -
serine/threonine protein kinase receptor
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
positive regulation of protein phosphorylation
protein phosphorylation
transmembrane receptor protein serine/threonine kinase signaling pathway
spermatogenesis
determination of left/right symmetry
mesoderm development
male gonad development
anterior/posterior pattern specification
regulation of signal transduction
positive regulation of pathway-restricted SMAD protein phosphorylation
phosphorylation
positive regulation of bone mineralization
BMP signaling pathway
regulation of BMP signaling pathway
activin receptor signaling pathway
positive regulation of activin receptor signaling pathway
sperm ejaculation
penile erection
positive regulation of erythrocyte differentiation
positive regulation of osteoblast differentiation
positive regulation of follicle-stimulating hormone secretion
embryonic skeletal system development
regulation of nitric-oxide synthase activity
Sertoli cell proliferation
cellular response to BMP stimulus
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
coreceptor activity
kinase activity
activin receptor activity, type II
transferase activity
transferase activity, transferring phosphorus-containing groups
activin-activated receptor activity
growth factor binding
PDZ domain binding
inhibin binding
protein self-association
metal ion binding
activin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0