Search
Search

Angpt1| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286062.1 | NP_001272991.1 | 4 | 1185 | ||
| NM_009640.4 | NP_033770.2 | 4 | 1185 | |||
| GenBank mRNA | AK012027.1 | - | 4 | 1121 | ||
| AK088439.1 | - | 4 | 1186 | |||
| AK172868.1 | - | 4 | 1005 | |||
| BC067410.1 | - | 4 | 1003 | 30436666 | ||
| U83509.1 | - | 4 | 975 | |||