Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCKL1 SIRNA Assays ›
          Gene Symbol:
          Uckl1
          Gene Name:
          uridine-cytidine kinase 1-like 1
          Gene Aliases:
          1110007H10Rik, Urkl1
          Chromosome Location:
          Chr.2: 181569149 - 181581973 on Build GRCm38
          UniGene ID:
          Mm.232400
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 163085

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Uckl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_026765.3 NP_081041.2 10 1042
          GenBank mRNA BC016535.1 - 10 1042 2655873

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metabolic process
          phosphorylation
          nucleotide binding
          uridine kinase activity
          ATP binding
          phosphofructokinase activity
          phosphorylase kinase regulator activity
          cobinamide kinase activity
          phytol kinase activity
          kinase activity
          phosphatidylinositol phosphate kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          transferase activity
          phenol kinase activity
          cyclin-dependent protein kinase activating kinase regulator activity
          inositol tetrakisphosphate 2-kinase activity
          heptose 7-phosphate kinase activity
          aminoglycoside phosphotransferase activity
          phosphatidylinositol 3-kinase activity
          eukaryotic elongation factor-2 kinase regulator activity
          eukaryotic elongation factor-2 kinase activator activity
          LPPG:FO 2-phospho-L-lactate transferase activity
          cytidine kinase activity
          glycerate 2-kinase activity
          (S)-lactate 2-kinase activity
          phosphoserine:homoserine phosphotransferase activity
          L-seryl-tRNA(Sec) kinase activity
          phosphocholine transferase activity
          polynucleotide 5'-hydroxyl-kinase activity
          ATP-dependent polynucleotide kinase activity
          GTP-dependent polynucleotide kinase activity
          farnesol kinase activity
          CTP:2-trans,-6-trans-farnesol kinase activity
          geraniol kinase activity
          geranylgeraniol kinase activity
          CTP:geranylgeraniol kinase activity
          prenol kinase activity
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol bisphosphate kinase activity
          inositol-3,4,6-trisphosphate 1-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0