Search
Search

Kank2| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145611.4 | NP_663586.3 | 3, 4 | 1417 | ||
| GenBank mRNA | AK031324.1 | - | 3, 4 | 1410 | ||
| AK045492.1 | - | 3, 4 | 1389 | |||
| AK049288.1 | - | 3, 4 | 1418 | |||
| BC006647.1 | - | 3, 4 | 1405 | |||
| BC010245.1 | - | 3, 4 | 1374 | 3710765 | ||