Search
Search

Eda2r| Gene Symbol: Eda2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161432.1 | NP_001154904.1 | 8 | 1669 | ||
| NM_175540.5 | NP_780749.3 | 8 | 1624 | |||
| GenBank mRNA | AK029268.1 | - | 1 | 398 | ||
| AK034909.1 | - | 8 | 1588 | |||