Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FN1 SIRNA Assays ›
          Gene Symbol:
          Fn1
          Gene Name:
          fibronectin 1
          Gene Aliases:
          E330027I09, Fn, Fn-1
          Chromosome Location:
          Chr.1: 71585473 - 71653234 on Build GRCm38
          UniGene ID:
          Mm.193099
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 165825

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fn1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276408.1 NP_001263337.1 17, 18 2775
          NM_001276409.1 NP_001263338.1 17, 18 2775
          NM_001276410.1 NP_001263339.1 17, 18 2775
          NM_001276411.1 NP_001263340.1 17, 18 2775
          NM_001276412.1 NP_001263341.1 17, 18 2775
          NM_001276413.1 NP_001263342.1 17, 18 2775
          NM_010233.2 NP_034363.1 17, 18 2775
          GenBank mRNA AK090130.1 - 17, 18 2720
          AK090135.1 - 17, 18 2716
          AK147249.1 - 8, 9 1125
          AK147315.1 - 17, 18 2714
          AK147639.1 - 17, 18 2714
          AK147654.1 - 17, 18 2714
          AK147663.1 - 17, 18 2714
          AK147683.1 - 17, 18 2714
          AK147701.1 - 17, 18 2713
          AK170654.1 - 17, 18 2714
          AK170755.1 - 17, 18 2743
          AK171339.1 - 17, 18 2714
          BC051082.1 - 17, 18 2684
          BC138421.1 - 17, 18 2639 8861441
          BC145271.1 - 17, 18 2639 9053825
          CN538894.1 - 3, 4 251

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell activation
          acute-phase response
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          glial cell migration
          regulation of cell shape
          positive regulation of peptidase activity
          peptide cross-linking
          extracellular matrix organization
          positive regulation of cell migration
          integrin activation
          substrate adhesion-dependent cell spreading
          endodermal cell differentiation
          wound healing
          negative regulation of apoptotic process
          positive regulation of axon extension
          positive regulation of chemotaxis
          cellular response to mercury ion
          cellular response to interleukin-1
          protease binding
          integrin binding
          protein binding
          heparin binding
          peptidase activator activity
          mercury ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0