Search
Search

Gk2| Gene Symbol: Gk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010294.1 | NP_034424.1 | 1 | 298 | ||
| GenBank mRNA | AF117734.1 | - | 1 | 298 | ||
| AK075761.1 | - | 1 | 211 | |||
| BC050764.1 | - | 1 | 311 | 6774427 | ||
| BC061147.1 | - | 1 | 277 | 6770827 | ||