Search
Search

Fcho2| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172591.3 | NP_766179.2 | 4 | 667 | ||
| GenBank mRNA | AK031041.1 | - | 4 | 670 | ||
| AK142282.1 | - | 4 | 353 | |||
| AK146566.1 | - | 4 | 401 | |||
| AK164135.1 | - | 4 | 413 | |||
| BC019933.1 | - | 5 | 403 | |||
| BC047122.1 | - | 5 | 403 | |||
| BC052456.1 | - | 4 | 364 | 6830607 | ||
| BC053718.1 | - | 4 | 383 | |||