Search
Search

SNAI1| Gene Symbol: SNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005985.3 | NP_005976.2 | 1, 2 | 161 | ||
| GenBank mRNA | AF125377.1 | - | 1, 2 | 138 | ||
| AF131208.1 | - | 1, 2 | 77 | |||
| AK313228.1 | - | 1, 2 | 157 | |||
| BC012910.1 | - | 1, 2 | 142 | 4537122 | ||
| BI822180.1 | - | 1, 2 | 148 | |||
| DA972913.1 | - | 1, 2 | 161 | |||