Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMK2D SIRNA Assays ›
          Gene Symbol:
          Camk2d
          Gene Name:
          calcium/calmodulin-dependent protein kinase II, delta
          Gene Aliases:
          2810011D23Rik, 8030469K03Rik, CaMK II, [d]-CaMKII
          Chromosome Location:
          Chr.3: 126596358 - 126846326 on Build GRCm38
          UniGene ID:
          Mm.255822
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 171960

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Camk2d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025438.2 NP_001020609.1 9, 10 1178
          NM_001025439.2 NP_001020610.1 9, 10 1178
          NM_001293663.1 NP_001280592.1 9, 10 1178
          NM_001293664.1 NP_001280593.1 9, 10 1178
          NM_001293665.1 NP_001280594.1 10, 11 1171
          NM_001293666.1 NP_001280595.1 9, 10 1178
          NM_023813.4 NP_076302.1 9, 10 1178
          GenBank mRNA AK012702.1 - 9, 10 799
          AK032524.1 - 9, 10 1125
          AK039076.1 - 9, 10 787
          AK044872.1 - 10, 11 1175
          AK082124.1 - 2, 3 113
          AK142435.1 - 9, 10 1121
          AK148840.1 - 9, 10 885
          AK220516.1 - 9, 10 1178
          BC042895.1 - 9, 10 1719
          BC057103.1 - 10, 11 1087

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          response to hypoxia
          regulation of sodium ion transport
          regulation of membrane depolarization
          protein phosphorylation
          calcium ion transport
          regulation of G2/M transition of mitotic cell cycle
          positive regulation of cardiac muscle hypertrophy
          regulation of cell communication by electrical coupling
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          positive regulation of smooth muscle cell migration
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cellular potassium ion homeostasis
          endoplasmic reticulum calcium ion homeostasis
          positive regulation of Rac protein signal transduction
          protein autophosphorylation
          positive regulation of smooth muscle cell proliferation
          protein oligomerization
          relaxation of cardiac muscle
          cardiac muscle contraction
          regulation of cellular localization
          cell growth involved in cardiac muscle cell development
          positive regulation of ERK1 and ERK2 cascade
          cardiac muscle cell contraction
          regulation of cardiac muscle cell action potential
          regulation of relaxation of cardiac muscle
          negative regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          negative regulation of sodium ion transmembrane transporter activity
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          protein binding
          calmodulin binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          sodium channel inhibitor activity
          titin binding
          protein homodimerization activity
          ion channel binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0