Search
Search

Rcc2| Gene Symbol: Rcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173867.5 | NP_776292.1 | 3 | 545 | ||
| GenBank mRNA | AK076040.1 | - | 3 | 476 | ||
| AK076046.1 | - | 776 | ||||
| AK129370.1 | - | 2 | 249 | |||
| BC020385.1 | - | 1 | 78 | |||
| BC060624.1 | - | 3 | 499 | |||
| BC086666.1 | - | 3 | 468 | 30634817 | ||
| CA751510.1 | - | 3 | 544 | |||