Search
Search

Rcc2| Gene Symbol: Rcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173867.5 | NP_776292.1 | 13 | 1825 | ||
| GenBank mRNA | AK076040.1 | - | 13 | 1756 | ||
| AK076046.1 | - | 2056 | ||||
| AK129370.1 | - | 12 | 1529 | |||
| BC020385.1 | - | 11 | 1358 | |||
| BC034759.1 | - | 10 | 1145 | |||
| BC060624.1 | - | 13 | 1778 | |||
| BC086666.1 | - | 13 | 1748 | 30634817 | ||