Search
Search

Ints5| Gene Symbol: Ints5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176843.3 | NP_789813.1 | 2 | 315 | ||
| GenBank mRNA | AK143868.1 | - | 2 | 277 | ||
| AK144770.1 | - | 2 | 290 | |||
| AK155499.1 | - | 2 | 277 | |||
| AK166149.1 | - | 2 | 288 | |||
| AK168795.1 | - | 2 | 288 | |||
| AK170584.1 | - | 2 | 277 | |||
| AK172260.1 | - | 2 | 277 | |||
| BC039214.1 | - | 2 | 297 | 4191047 | ||