Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOL3 SIRNA Assays ›
Gene Symbol:
Nol3
Gene Name:
nucleolar protein 3 (apoptosis repressor with CARD domain)
Gene Aliases:
ARC, B430311C09Rik, MYC, NOP, Nop30
Chromosome Location:
Chr.8: 105275937 - 105281939 on Build GRCm38
UniGene ID:
Mm.475715
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
174808
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nol3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_030152.4
NP_084428.1
4
1591
GenBank mRNA
AK053846.1
-
4
1592
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
blood vessel remodeling
response to ischemia
mRNA splice site selection
mRNA processing
regulation of gene expression
cardiac muscle cell apoptotic process
negative regulation of striated muscle cell apoptotic process
negative regulation of cardiac muscle cell apoptotic process
negative regulation of tumor necrosis factor-mediated signaling pathway
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
negative regulation of muscle atrophy
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
response to injury involved in regulation of muscle adaptation
regulation of NF-kappaB import into nucleus
regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
myoblast differentiation
smooth muscle cell proliferation
protein oligomerization
negative regulation of cytosolic calcium ion concentration
negative regulation of mitochondrial calcium ion concentration
negative regulation of necrotic cell death
cellular response to hypoxia
negative regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of mitochondrial membrane permeability involved in apoptotic process
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
negative regulation of death-inducing signaling complex assembly
negative regulation of hydrogen peroxide-induced cell death
negative regulation of protein targeting to mitochondrion
negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
receptor binding
death receptor binding
calcium ion binding
protein binding
kinase binding
phosphatase binding
death effector domain binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
metal ion binding
caspase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0