Search
Search

Rab2b| Gene Symbol: Rab2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172601.3 | NP_766189.1 | 8 | 883 | ||
| GenBank mRNA | AK038393.1 | - | 8 | 818 | ||
| AK044202.1 | - | 8 | 760 | |||
| AK160249.1 | - | 8 | 1034 | |||
| AK160351.1 | - | 8 | 749 | |||
| AK169657.1 | - | 7 | 758 | |||
| BC033312.1 | - | 8 | 780 | 4481516 | ||
| BC046334.1 | - | 8 | 825 | 6492838 | ||
| BC054719.1 | - | 8 | 816 | |||
| BC061513.1 | - | 3 | 1713 | |||