Search
Search

Cul5| Gene Symbol: Cul5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161618.1 | NP_001155090.1 | 9, 10 | 1486 | ||
| NM_027807.3 | NP_082083.2 | 10, 11 | 1567 | |||
| GenBank mRNA | AK014894.1 | - | 10, 11 | 1160 | ||
| AK031955.1 | - | 10, 11 | 1570 | |||
| AK046030.1 | - | 10, 11 | 1538 | |||
| AK162082.1 | - | 1490 | ||||
| BC075710.1 | - | 10, 11 | 1496 | 6312416 | ||