Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRFIP2 SIRNA Assays ›
          Gene Symbol:
          LRRFIP2
          Gene Name:
          LRR binding FLII interacting protein 2
          Gene Aliases:
          HUFI-2
          Chromosome Location:
          Chr.3: 37052626 - 37176817 on Build GRCh38
          UniGene ID:
          Hs.740522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17849

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRFIP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134369.2 NP_001127841.1 14 1511
          NM_001282691.1 NP_001269620.1 18 1500
          NM_006309.3 NP_006300.1 27 2101
          NM_017724.2 NP_060194.1 13 1439
          XM_005265539.2 XP_005265596.1 28 3101
          XM_005265540.1 XP_005265597.1 29 2384
          XM_005265550.1 XP_005265607.1 20 1937
          XM_005265551.1 XP_005265608.1 19 1826
          XM_005265553.1 XP_005265610.1 16 1673
          XM_005265554.2 XP_005265611.1 16 2405
          XM_005265556.1 XP_005265613.1 15 1601
          XM_006713385.1 XP_006713448.1 28 2366
          XM_006713386.1 XP_006713449.1 27 2327
          XM_006713387.1 XP_006713450.1 27 2324
          XM_006713388.1 XP_006713451.1 27 2321
          XM_006713389.1 XP_006713452.1 27 2318
          XM_006713390.1 XP_006713453.1 28 2361
          XM_006713392.1 XP_006713455.1 27 2297
          XM_006713393.1 XP_006713456.1 27 2279
          XM_006713394.1 XP_006713457.1 26 2276
          XM_006713395.1 XP_006713458.1 27 2276
          XM_006713396.1 XP_006713459.1 26 2261
          XM_006713397.1 XP_006713460.1 25 2153
          XM_006713400.1 XP_006713463.1 17 1718
          XM_006713404.1 XP_006713467.1 14 1451
          XM_011534217.1 XP_011532519.1 28 2369
          XM_011534218.1 XP_011532520.1 29 2412
          XM_011534219.1 XP_011532521.1 29 2480
          XM_011534221.1 XP_011532523.1 26 2261
          XM_011534222.1 XP_011532524.1 26 2174
          XM_011534229.1 XP_011532531.1 18 1761
          XM_017007465.1 XP_016862954.1 25 2216
          XM_017007466.1 XP_016862955.1 26 2210
          XM_017007467.1 XP_016862956.1 26 2204
          XM_017007468.1 XP_016862957.1 23 2108
          XM_017007469.1 XP_016862958.1 25 2102
          XM_017007470.1 XP_016862959.1 23 2066
          XM_017007471.1 XP_016862960.1 18 1775
          XM_017007472.1 XP_016862961.1 18 1733
          XM_017007473.1 XP_016862962.1 17 1703
          XM_017007474.1 XP_016862963.1 17 1682
          XM_017007475.1 XP_016862964.1 17 2414
          XM_017007476.1 XP_016862965.1 17 1716
          XM_017007477.1 XP_016862966.1 16 2342
          XM_017007478.1 XP_016862967.1 15 1580
          XM_017007479.1 XP_016862968.1 16 1580
          XM_017007480.1 XP_016862969.1 15 1824
          XM_017007481.1 XP_016862970.1 15 1571
          XM_017007482.1 XP_016862971.1 14 1508
          XM_017007483.1 XP_016862972.1 14 2210
          XM_017007484.1 XP_016862973.1 15 1521
          GenBank mRNA AF115509.1 - 27 2060
          AK000255.1 - 12 1124
          AK024692.1 - 6 457
          AK124432.1 - 5 3235
          AK291514.1 - 14 1488
          AK302282.1 - 18 1474
          BC053668.1 - 13 1396 6146978
          BX648794.1 - 10 890
          CR749705.1 - 4 1828

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          biological_process
          Wnt signaling pathway
          protein binding
          LRR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0