Search
Search

Cpa5| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144537.3 | NP_653120.1 | 10 | 1552 | ||
| GenBank mRNA | AF466283.1 | - | 10 | 1553 | ||
| AK015256.1 | - | 10 | 1555 | |||
| BC138114.1 | - | 9 | 1070 | 8861133 | ||
| BC145151.1 | - | 9 | 937 | |||