Search
Search

Rps27l| Gene Symbol: Rps27l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026467.3 | NP_080743.1 | 4 | 311 | ||
| GenBank mRNA | AK007646.1 | - | 1 | 18 | ||
| AK007689.1 | - | 4 | 312 | |||
| AK011262.1 | - | 4 | 330 | |||
| AK151083.1 | - | 4 | 330 | |||
| BC058115.1 | - | 4 | 312 | 6407738 | ||