Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CELF1 SIRNA Assays ›
          Gene Symbol:
          CELF1
          Gene Name:
          CUGBP, Elav-like family member 1
          Gene Aliases:
          BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
          Chromosome Location:
          Chr.11: 47465937 - 47565569 on Build GRCh38
          UniGene ID:
          Hs.595333
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18094

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CELF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025596.2 NP_001020767.1 1 164
          NM_001172639.1 NP_001166110.1 4 1129
          NM_001172640.1 NP_001166111.1 1 164
          NM_006560.3 NP_006551.1 2 413
          NM_198700.2 NP_941989.1 2 355
          XM_011519848.2 XP_011518150.1 2 236
          XM_011519850.2 XP_011518152.2 2 475
          XM_011519852.2 XP_011518154.1 4 584
          XM_011519854.1 XP_011518156.1 6 692
          XM_011519855.1 XP_011518157.1 2 413
          XM_011519856.1 XP_011518158.1 3 494
          XM_011519857.1 XP_011518159.1 2 365
          XM_011519858.1 XP_011518160.1 2 460
          XM_011519859.1 XP_011518161.1 6 692
          XM_017017101.1 XP_016872590.1 5 576
          XM_017017102.1 XP_016872591.1 4 495
          XM_017017103.1 XP_016872592.1 3 388
          XM_017017104.1 XP_016872593.1 3 388
          XM_017017105.1 XP_016872594.1 3 388
          XM_017017106.1 XP_016872595.1 5 576
          XM_017017107.1 XP_016872596.1 3 388
          XM_017017108.1 XP_016872597.1 3 388
          XM_017017109.1 XP_016872598.1 2 475
          XM_017017110.1 XP_016872599.1 3 388
          XM_017017111.1 XP_016872600.1 3 388
          XM_017017112.1 XP_016872601.1 3 388
          XM_017017113.1 XP_016872602.1 3 388
          XM_017017114.1 XP_016872603.1 3 388
          XM_017017115.1 XP_016872604.1 4 416
          XM_017017116.1 XP_016872605.1 3 451
          XM_017017117.1 XP_016872606.1 2 212
          XM_017017118.1 XP_016872607.1 2 334
          XM_017017119.1 XP_016872608.1 4 553
          XM_017017120.1 XP_016872609.1 2 365
          XM_017017121.1 XP_016872610.1 2 460
          XM_017017122.1 XP_016872611.1 2 365
          XM_017017123.1 XP_016872612.1 2 413
          XM_017017124.1 XP_016872613.1 4 507
          XM_017017125.1 XP_016872614.1 2 236
          XM_017017126.1 XP_016872615.1 2 365
          XM_017017127.1 XP_016872616.1 2 389
          XM_017017128.1 XP_016872617.1 2 413
          XM_017017129.1 XP_016872618.1 3 411
          XM_017017130.1 XP_016872619.1 2 365
          XM_017017131.1 XP_016872620.1 2 389
          XM_017017132.1 XP_016872621.1 2 413
          XM_017017133.1 XP_016872622.1 2 204
          XM_017017134.1 XP_016872623.1 3 343
          XM_017017135.1 XP_016872624.1 2 365
          GenBank mRNA AB210019.1 - 4 1129
          AF248648.1 - 1 172
          AF267533.1 - 1 165
          AF267534.1 - 1 165
          AJ007988.1 - 1 154
          AK304430.1 - 1 107
          BC031079.1 - 2 355 5297141
          CB960956.1 - 3 316
          DA217269.1 - 4 436
          DA320861.1 - 2 414
          DB162286.1 - 2 313
          U63289.1 - 2 291

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001025421, XM_006234522, XM_006234520, XM_006234526, XM_006234516, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001244891, NM_017368, NM_001244903, NM_198683, BU056861, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splice site selection
          mRNA processing
          germ cell development
          spermatid development
          embryo development
          RNA interference
          negative regulation of translation
          positive regulation of multicellular organism growth
          regulation of RNA splicing
          nucleotide binding
          translation repressor activity, nucleic acid binding
          RNA binding
          mRNA binding
          protein binding
          translation initiation factor binding
          BRE binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0