Search
Search

Mtcl1| Gene Symbol: Mtcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114098.1 | NP_001107570.1 | 13 | 4675 | ||
| NM_172963.4 | NP_766551.3 | 13 | 4673 | |||
| GenBank mRNA | AK122373.1 | - | 2 | 186 | ||
| AK147205.1 | - | 13 | 4675 | |||
| AK147631.1 | - | 13 | 4667 | |||
| AK147691.1 | - | 13 | 4667 | |||
| BC032156.2 | - | 9 | 1795 | |||
| BC060225.1 | - | 11 | 2921 | 30356997 | ||