Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FRS2 SIRNA Assays ›
          Gene Symbol:
          FRS2
          Gene Name:
          fibroblast growth factor receptor substrate 2
          Gene Aliases:
          FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
          Chromosome Location:
          Chr.12: 69470349 - 69579793 on Build GRCh38
          UniGene ID:
          Hs.593446
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18238

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FRS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042555.2 NP_001036020.1 6, 7 577
          NM_001278351.1 NP_001265280.1 7, 8 698
          NM_001278353.1 NP_001265282.1 7, 8 641
          NM_001278354.1 NP_001265283.1 6, 7 630
          NM_001278355.1 NP_001265284.1 6, 7 612
          NM_001278356.1 NP_001265285.1 5, 6 516
          NM_001278357.1 NP_001265286.1 3, 4 377
          NM_006654.4 NP_006645.3 6, 7 584
          XM_017018717.1 XP_016874206.1 7, 8 664
          XM_017018718.1 XP_016874207.1 7, 8 658
          XM_017018719.1 XP_016874208.1 2, 3 123
          GenBank mRNA AF036717.1 - 1, 2 79
          AK312477.1 - 5, 6 435
          BC021562.1 - 3, 4 320 4556225
          BP217337.1 - 6, 7 544
          BP230333.1 - 6, 7 550
          BP280775.1 - 6, 7 542
          BP311537.1 - 5, 6 483
          BP363404.1 - 5, 6 484
          BU166503.1 - 7, 8 582
          DA189114.1 - 5, 6 488
          DA721439.1 - 6, 7 559

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_177798, BC043109, AK028813, BC055334.

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          activation of MAPK activity
          gastrulation with mouth forming second
          organ induction
          ventricular septum development
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          G-protein coupled receptor signaling pathway
          neuroblast proliferation
          fibroblast growth factor receptor signaling pathway
          anterior/posterior axis specification, embryo
          regulation of phosphatidylinositol 3-kinase signaling
          forebrain development
          phosphatidylinositol-3-phosphate biosynthetic process
          regulation of apoptotic process
          positive regulation of GTPase activity
          optic placode formation involved in camera-type eye formation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          regulation of epithelial cell proliferation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          lens fiber cell development
          regulation of ERK1 and ERK2 cascade
          transmembrane receptor protein tyrosine kinase adaptor activity
          Ras guanyl-nucleotide exchange factor activity
          fibroblast growth factor receptor binding
          insulin receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          1-phosphatidylinositol-3-kinase activity
          phosphatase activator activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-sdnzb:80/100.66.78.42:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0