Search
Search

Rfx5| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017395.2 | NP_059091.2 | 4 | 278 | ||
| GenBank mRNA | AB221679.1 | - | 4 | 259 | ||
| AF209854.1 | - | 4 | 259 | |||
| AK088686.1 | - | 5 | 330 | |||
| BB666132.1 | - | 5 | 304 | |||
| BB854843.1 | - | 5 | 371 | |||
| BC051965.1 | - | 5 | 315 | 5702640 | ||
| BC057336.1 | - | 5 | 304 | |||
| BC075717.1 | - | 6 | 387 | |||
| BC118948.2 | - | 4 | 259 | 40092396 | ||
| BC118949.2 | - | 4 | 259 | 40092397 | ||
| BY020562.1 | - | 5 | 340 | |||
| BY230827.1 | - | 5 | 300 | |||