Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTF2IRD1 SIRNA Assays ›
Gene Symbol:
Gtf2ird1
Gene Name:
general transcription factor II I repeat domain-containing 1
Gene Aliases:
1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320
Chromosome Location:
Chr.5: 134357661 - 134456716 on Build GRCm38
UniGene ID:
Mm.332735
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
185993
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gtf2ird1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081462.2
NP_001074931.1
9
1419
NM_001081463.2
NP_001074932.1
9
1419
NM_001081464.2
NP_001074933.1
9
1419
NM_001081465.1
NP_001074934.1
9
1362
NM_001081466.2
NP_001074935.1
9
1419
NM_001081467.2
NP_001074936.1
9
1419
NM_001081468.2
NP_001074937.1
9
1419
NM_001081469.2
NP_001074938.1
9
1419
NM_001081470.2
NP_001074939.1
9
1419
NM_001244936.1
NP_001231865.1
9
1419
NM_020331.3
NP_065064.2
9
1419
GenBank mRNA
AF257475.1
-
9
1395
AF260133.1
-
9
1390
AF343348.1
-
9
1287
AF343349.1
-
9
1293
AF343350.1
-
9
1289
AF343351.1
-
9
1287
AF497637.1
-
9
1279
AF497638.1
-
9
1279
AF497639.1
-
9
1279
AF497640.1
-
9
1287
AF497641.1
-
9
1293
AF497642.1
-
9
1289
AK078488.1
-
9
1287
AK079487.1
-
9
1359
AK147196.1
-
9
1402
AY030287.1
-
9
1390
AY030288.1
-
9
1390
AY030289.1
-
9
1390
BC050822.1
-
9
1414
6393328
BC057091.1
-
9
1241
5703300
BC058964.1
-
9
1241
5703300
BC083150.1
-
9
1321
30459984
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
embryo development
transition between slow and fast fiber
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0