Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YAP1 SIRNA Assays ›
          Gene Symbol:
          Yap1
          Gene Name:
          yes-associated protein 1
          Gene Aliases:
          AI325207, Yap, Yap65, Yki, Yorkie
          Chromosome Location:
          Chr.9: 7931999 - 8004596 on Build GRCm38
          UniGene ID:
          Mm.221992
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 187077

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Yap1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171147.1 NP_001164618.1 9 1662
          NM_009534.3 NP_033560.1 8 1614
          GenBank mRNA AK160543.1 - 2 318
          BC014733.1 - 9 1659 4239820
          BC039125.1 - 8 1469 2649070
          BC094313.1 - 8 1602 5697882
          X80508.1 - 8 1606

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001034002, XM_006242489, XM_006242490, XM_008765894, XM_006242492, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          vasculogenesis
          embryonic heart tube morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          cell proliferation
          positive regulation of cell proliferation
          regulation of keratinocyte proliferation
          keratinocyte differentiation
          negative regulation of epithelial cell differentiation
          notochord development
          somatic stem cell population maintenance
          hippo signaling
          regulation of cell proliferation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of organ growth
          paraxial mesoderm development
          lateral mesoderm development
          regulation of neurogenesis
          contact inhibition
          bud elongation involved in lung branching
          lung epithelial cell differentiation
          regulation of canonical Wnt signaling pathway
          cellular response to retinoic acid
          cellular response to gamma radiation
          regulation of stem cell proliferation
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of canonical Wnt signaling pathway
          positive regulation of stem cell population maintenance
          negative regulation of nucleic acid-templated transcription
          negative regulation of stem cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          protein C-terminus binding
          transcription regulatory region DNA binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0