Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LILRA6 SIRNA Assays ›
          Gene Symbol:
          LILRA6 LILRB3
          Gene Name:
          leukocyte immunoglobulin like receptor A6
          leukocyte immunoglobulin like receptor B3
          Gene Aliases:
          CD85b, ILT-8, ILT5, ILT8, LILRB3, LILRB6
          CD85A, HL9, ILT-5, ILT5, LILRA6, LIR-3, LIR3, PIR-B, PIRB
          Chromosome Location:
          Chr.19: 54236592 - 54242768 on Build GRCh38
          Chromosome Location:
          Chr.19: 54216278 - 54223087 on Build GRCh38
          UniGene ID:
          Hs.688335
          Hs.631592
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 18750

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LILRA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA KC918856.1 - 8, 9 1470
          KC918859.1 - 9, 10 1470
          Gene Symbol: LILRB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081450.2 NP_001074919.2 8, 9 1537
          NM_001320960.1 NP_001307889.1 9, 10 1588
          NM_006864.3 NP_006855.3 8, 9 1537
          NR_135494.1 8, 9 1451
          NR_135495.1 8, 9 1457
          NR_135496.1 9, 10 1620
          XM_011526381.2 XP_011524683.1 9, 10 1588
          XM_011526382.2 XP_011524684.1 9, 10 1588
          XM_017026218.1 XP_016881707.1 8, 9 1456
          XM_017026219.1 XP_016881708.1 8, 9 1456
          XM_017026220.1 XP_016881709.1 8, 9 1456
          XM_017026221.1 XP_016881710.1 8, 9 1456
          XM_017026222.1 XP_016881711.1 7, 8 1405
          XM_017026223.1 XP_016881712.1 7, 8 1405
          GenBank mRNA AF000575.1 - 8, 9 1426
          AF009632.1 - 8, 9 1428
          AF009633.1 - 8, 9 1428
          AF009635.1 - 8, 9 1428
          AF009637.1 - 9, 10 1479
          AF009638.1 - 8, 9 1428
          AF009639.1 - 8, 9 1428
          AF009641.1 - 8, 9 1428
          AF009642.1 - 8, 9 1428
          AF009643.1 - 8, 9 1342
          AF009644.1 - 8, 9 1348
          AF025533.1 - 8, 9 1492
          AF031553.1 - 8, 9 1428
          AF031554.1 - 8, 9 1428
          AF031555.1 - 8, 9 1428
          AF031556.1 - 8, 9 1428
          AK223265.1 - 8, 9 1479
          AK290917.1 - 8, 9 1507
          BC028153.1 - 9, 10 1511
          BC104993.1 - 8, 9 1445 8143996
          BC112198.1 - 8, 9 1445 8327666
          EU814627.1 - 8, 9 1426
          EU814628.1 - 8, 9 1426
          KC918852.1 - 8, 9 1470
          KC918854.1 - 8, 9 1470
          KC918855.1 - 8, 9 1470
          KC918857.1 - 8, 9 1470
          KC918858.1 - 8, 9 1470
          KC918860.1 - 8, 9 1470
          KC918863.1 - 8, 9 1470
          U91928.1 - 8, 9 1475

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          defense response
          cell surface receptor signaling pathway
          negative regulation of osteoclast differentiation
          receptor activity
          transmembrane signaling receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0