Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FES SIRNA Assays ›
          Gene Symbol:
          FES
          Gene Name:
          FES proto-oncogene, tyrosine kinase
          Gene Aliases:
          FPS
          Chromosome Location:
          Chr.15: 90884421 - 90895776 on Build GRCh38
          UniGene ID:
          Hs.7636
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 189

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FES
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143783.1 NP_001137255.1 10 1389
          NM_001143784.1 NP_001137256.1 10 1353
          NM_001143785.1 NP_001137257.1 10 1298
          NM_002005.3 NP_001996.1 12 1659
          XM_005254880.1 XP_005254937.1 11 1474
          XM_005254882.1 XP_005254939.1 11 1465
          XM_017022005.1 XP_016877494.1 12 1643
          XM_017022006.1 XP_016877495.1 11 1516
          XM_017022007.1 XP_016877496.1 11 1433
          XM_017022008.1 XP_016877497.1 10 1307
          XM_017022009.1 XP_016877498.1 12 1740
          XM_017022010.1 XP_016877499.1 11 1591
          GenBank mRNA AK300595.1 - 10 1288
          AK300605.1 - 12 1617
          AK312545.1 - 12 1648
          AY513654.1 - 9 1179
          AY513655.1 - 9 1179
          AY513656.1 - 10 1389
          AY513657.1 - 10 1353
          BC035357.1 - 12 1635 5170548
          X52192.1 - 12 1638

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          protein phosphorylation
          chemotaxis
          epidermal growth factor receptor signaling pathway
          multicellular organism development
          cell proliferation
          regulation of cell shape
          positive regulation of neuron projection development
          peptidyl-tyrosine phosphorylation
          regulation of cell adhesion
          positive regulation of microtubule polymerization
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          regulation of mast cell degranulation
          innate immune response
          regulation of cell differentiation
          positive regulation of myeloid cell differentiation
          protein autophosphorylation
          centrosome organization
          regulation of vesicle-mediated transport
          regulation of cell motility
          positive regulation of actin cytoskeleton reorganization
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          microtubule binding
          immunoglobulin receptor binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0