Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYC SIRNA Assays ›
          Gene Symbol:
          Myc
          Gene Name:
          myelocytomatosis oncogene
          Gene Aliases:
          RNCMYC, c-myc, mMyc
          Chromosome Location:
          Chr.7: 102586313 - 102591240 on Build Rnor_6.0
          UniGene ID:
          Rn.12072
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 189043

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Myc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012603.2 NP_036735.2 2, 3 1335
          GenBank mRNA AY679729.1 - 2, 3 1172
          AY679730.1 - 2, 3 1335
          BC091699.1 - 2, 3 1141 7303974

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          B cell apoptotic process
          positive regulation of mesenchymal cell proliferation
          regulation of oxidative phosphorylation
          positive regulation of B cell apoptotic process
          glucose metabolic process
          chromatin remodeling
          transcription, DNA-templated
          DNA-templated transcription, initiation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          pyruvate transport
          amino acid transport
          cellular iron ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          inner mitochondrial membrane organization
          cell cycle arrest
          regulation of mitotic cell cycle
          cell proliferation
          positive regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          glycoprotein metabolic process
          response to radiation
          pathogenesis
          response to wounding
          flavonoid metabolic process
          response to gamma radiation
          regulation of gene expression
          negative regulation of gene expression
          positive regulation of mitochondrial membrane potential
          Wnt signaling pathway
          protein processing
          transformation of host cell by virus
          hypothalamus development
          negative regulation of protein binding
          regulation of telomere maintenance
          response to estradiol
          cellular response to UV
          cellular response to drug
          skeletal muscle cell differentiation
          middle ear morphogenesis
          response to drug
          regulation of apoptotic process
          positive regulation of catalytic activity
          response to alkaloid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          pigmentation
          canonical Wnt signaling pathway involved in positive regulation of wound healing
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          G0 to G1 transition
          negative regulation of monocyte differentiation
          positive regulation of cell cycle
          positive regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          lactic acid secretion
          positive regulation of fibroblast proliferation
          negative regulation of fibroblast proliferation
          skeletal system morphogenesis
          positive regulation of epithelial cell proliferation
          detection of mechanical stimulus involved in sensory perception of sound
          chromosome organization
          negative regulation of cell division
          canonical Wnt signaling pathway
          negative regulation of cell death
          cellular response to carbohydrate stimulus
          cellular response to interleukin-1
          cellular response to estrogen stimulus
          cellular response to organic cyclic compound
          cellular response to hydrostatic pressure
          fungal-type cell wall polysaccharide metabolic process
          positive regulation of metanephric cap mesenchymal cell proliferation
          positive regulation of cellular respiration
          positive regulation of DNA biosynthetic process
          positive regulation of response to DNA damage stimulus
          positive regulation of ATP biosynthetic process
          positive regulation of apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein complex binding
          sequence-specific DNA binding
          protein heterodimerization activity
          protein dimerization activity
          repressing transcription factor binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0