Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
189380
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
5
595
GenBank mRNA
BC062028.1
-
4
564
5623881
DN931898.1
-
4
613
FQ209537.1
-
5
595
FQ209603.1
-
5
595
FQ209620.1
-
4
580
FQ209649.1
-
5
595
FQ209757.1
-
5
596
FQ210412.1
-
5
595
FQ211152.1
-
5
595
FQ211263.1
-
5
594
FQ212554.1
-
5
595
FQ212743.1
-
5
595
FQ212875.1
-
5
595
FQ212927.1
-
5
596
FQ213085.1
-
5
595
FQ213427.1
-
5
595
FQ213510.1
-
5
595
FQ213712.1
-
5
595
FQ213847.1
-
5
596
FQ214553.1
-
5
596
FQ214850.1
-
5
595
FQ214997.1
-
5
595
FQ215007.1
-
5
595
FQ215548.1
-
5
597
FQ215662.1
-
5
595
FQ215682.1
-
5
595
FQ215870.1
-
5
595
FQ216120.1
-
5
596
FQ216296.1
-
5
595
FQ216389.1
-
5
596
FQ216406.1
-
5
598
FQ216495.1
-
5
595
FQ216618.1
-
5
595
FQ216655.1
-
5
596
FQ216733.1
-
5
597
FQ216774.1
-
5
595
FQ216813.1
-
5
594
FQ216851.1
-
5
595
FQ216913.1
-
5
596
FQ217406.1
-
5
595
FQ218561.1
-
5
595
FQ218832.1
-
5
595
FQ218943.1
-
5
595
FQ219025.1
-
5
598
FQ219184.1
-
5
599
FQ219389.1
-
5
595
FQ219457.1
-
5
595
FQ219678.1
-
5
595
FQ219817.1
-
5
595
FQ219837.1
-
5
595
FQ219855.1
-
5
595
FQ219923.1
-
5
595
FQ220008.1
-
5
595
FQ220051.1
-
5
595
FQ220077.1
-
5
595
FQ220115.1
-
5
595
FQ220181.1
-
5
595
FQ220201.1
-
5
593
FQ220213.1
-
5
595
FQ220247.1
-
5
596
FQ220282.1
-
5
597
FQ220284.1
-
5
595
FQ220302.1
-
5
595
FQ220338.1
-
5
595
FQ220345.1
-
5
596
FQ220370.1
-
5
595
FQ220406.1
-
5
595
FQ220413.1
-
5
595
FQ220420.1
-
5
596
FQ220433.1
-
5
592
FQ220444.1
-
5
595
FQ220445.1
-
5
591
FQ220449.1
-
5
595
FQ220500.1
-
5
595
FQ220514.1
-
5
595
FQ220527.1
-
5
595
FQ220533.1
-
5
627
FQ220542.1
-
5
595
FQ220545.1
-
5
595
FQ220634.1
-
5
595
FQ220654.1
-
5
595
FQ220663.1
-
5
595
FQ220684.1
-
5
596
FQ220690.1
-
5
597
FQ220769.1
-
5
595
FQ220787.1
-
5
595
FQ220801.1
-
5
595
FQ220833.1
-
5
595
FQ220838.1
-
5
595
FQ220888.1
-
5
595
FQ220932.1
-
5
596
FQ221146.1
-
5
595
FQ221380.1
-
5
595
FQ221533.1
-
5
596
FQ221556.1
-
5
593
FQ222171.1
-
5
595
FQ222181.1
-
5
595
FQ222327.1
-
5
595
FQ222772.1
-
5
595
FQ222906.1
-
5
595
FQ223121.1
-
5
595
FQ223487.1
-
5
595
FQ224547.1
-
5
595
FQ224834.1
-
5
595
FQ224846.1
-
5
595
FQ225049.1
-
5
595
FQ225170.1
-
5
595
FQ225608.1
-
5
595
FQ225654.1
-
5
597
FQ225860.1
-
5
597
FQ225886.1
-
5
595
FQ226306.1
-
5
597
FQ227381.1
-
5
595
FQ227522.1
-
5
590
FQ227719.1
-
5
595
FQ227837.1
-
5
595
FQ227937.1
-
5
596
FQ227941.1
-
5
595
FQ227942.1
-
5
715
FQ228548.1
-
5
595
FQ228995.1
-
5
595
FQ228996.1
-
5
595
FQ229139.1
-
5
597
FQ229159.1
-
5
595
FQ229164.1
-
5
596
FQ229208.1
-
5
595
FQ229241.1
-
5
595
FQ229244.1
-
5
595
FQ229248.1
-
3
475
FQ229272.1
-
5
595
FQ229273.1
-
5
596
FQ229303.1
-
5
595
FQ229337.1
-
5
595
FQ229352.1
-
5
595
FQ229365.1
-
5
595
FQ229397.1
-
5
595
FQ229408.1
-
5
596
FQ229433.1
-
5
595
FQ229448.1
-
5
595
FQ229521.1
-
5
595
FQ229547.1
-
5
595
FQ229554.1
-
5
595
FQ229566.1
-
5
591
FQ229610.1
-
5
595
FQ229626.1
-
5
592
FQ229726.1
-
5
595
FQ229740.1
-
5
595
FQ229742.1
-
5
595
FQ229776.1
-
5
595
FQ229808.1
-
5
595
FQ229815.1
-
5
595
FQ229822.1
-
5
595
FQ229830.1
-
5
597
FQ229883.1
-
5
597
FQ229901.1
-
5
595
FQ229929.1
-
5
595
FQ229952.1
-
5
595
FQ229974.1
-
5
595
FQ230005.1
-
5
595
FQ230055.1
-
5
594
FQ230127.1
-
5
595
FQ230267.1
-
5
598
FQ230943.1
-
5
595
FQ231445.1
-
5
597
FQ231525.1
-
5
595
FQ231591.1
-
5
597
FQ231710.1
-
5
596
FQ231733.1
-
5
596
FQ231897.1
-
5
597
FQ231967.1
-
5
595
FQ232029.1
-
5
596
FQ232154.1
-
5
595
FQ232165.1
-
5
595
FQ232310.1
-
5
594
FQ232319.1
-
5
596
FQ232697.1
-
5
597
FQ232767.1
-
5
595
FQ232951.1
-
5
595
FQ233152.1
-
5
594
FQ233377.1
-
5
595
FQ233473.1
-
5
595
FQ233732.1
-
5
595
FQ233737.1
-
5
595
FQ233808.1
-
5
595
FQ234195.1
-
5
595
FQ234341.1
-
5
595
FQ234613.1
-
5
595
FQ234733.1
-
5
595
FQ234735.1
-
5
485
FQ234863.1
-
5
595
FQ235063.1
-
5
595
FQ235079.1
-
5
595
FQ235122.1
-
5
595
FQ235259.1
-
5
595
X15096.1
-
5
595
Z29530.1
-
4
551
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0