Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HAND2 SIRNA Assays ›
Gene Symbol:
Hand2
Gene Name:
heart and neural crest derivatives expressed 2
Gene Aliases:
-
Chromosome Location:
Chr.16: 36370715 - 36373634 on Build Rnor_6.0
UniGene ID:
Rn.41057
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
189464
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hand2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022696.1
NP_073187.1
621
GenBank mRNA
Y08138.1
-
621
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_021973, BC101406, BC109383, AF087940, BF313018, and others.
This siRNA also targets the following transcripts in Mouse: NM_010402, AK035160, AJ131846.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
in utero embryonic development
heart looping
suckling behavior
heart morphogenesis
cardiac right ventricle formation
cardiac neural crest cell migration involved in outflow tract morphogenesis
regulation of secondary heart field cardioblast proliferation
apoptotic process involved in heart morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
adult heart development
mesenchymal cell proliferation
negative regulation of cardiac muscle cell apoptotic process
neural crest cell development
cell differentiation
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
tongue development
negative regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
sympathetic nervous system development
thymus development
peripheral nervous system neuron development
palate development
mesenchyme development
cartilage morphogenesis
coronary artery morphogenesis
visceral serous pericardium development
cardiac neural crest cell development involved in outflow tract morphogenesis
cell proliferation involved in outflow tract morphogenesis
determination of heart left/right asymmetry
cellular response to retinoic acid
cellular response to organic cyclic compound
positive regulation of transcription regulatory region DNA binding
positive regulation of transcription from RNA polymerase II promoter involved in norepinephrine biosynthetic process
positive regulation of semaphorin-plexin signaling pathway involved in outflow tract morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
AT DNA binding
transcription coactivator activity
protein binding
transcription factor binding
activating transcription factor binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein dimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0