Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL30 SIRNA Assays ›
          Gene Symbol:
          Rpl30 LOC100362027 LOC103690975 RGD1562394 Rpl30l1
          Gene Name:
          ribosomal protein L30
          ribosomal protein L30-like
          60S ribosomal protein L30-like
          similar to ribosomal protein L30
          ribosomal protein L30-like 1
          Gene Aliases:
          -
          -
          -
          -
          Gm7429, RGD1562397
          Chromosome Location:
          Chr.7: 73213538 - 73216431 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 38024456 - 38024944 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 42741210 - 42744274 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 22031576 - 22031980 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 95416242 - 95416632 on Build Rnor_6.0
          UniGene ID:
          Rn.161950
          -
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID 189468

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpl30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022699.3 NP_073190.1 3, 4 241
          GenBank mRNA BC058471.1 - 3, 4 245 6919868
          FQ210232.1 - 3, 4 238
          FQ210454.1 - 3, 4 237
          FQ210629.1 - 3, 4 241
          FQ211005.1 - 3, 4 243
          FQ211389.1 - 3, 4 239
          FQ211694.1 - 3, 4 238
          FQ211731.1 - 3, 4 237
          FQ211953.1 - 3, 4 241
          FQ214466.1 - 3, 4 238
          FQ217830.1 - 3, 4 238
          FQ217913.1 - 3, 4 238
          FQ221046.1 - 3, 4 238
          FQ221166.1 - 3, 4 238
          FQ221653.1 - 3, 4 246
          FQ221662.1 - 3, 4 239
          FQ221710.1 - 3, 4 238
          FQ221740.1 - 3, 4 237
          FQ221836.1 - 3, 4 238
          FQ222551.1 - 3, 4 238
          FQ222703.1 - 3, 4 237
          FQ223191.1 - 3, 4 237
          FQ223319.1 - 3, 4 238
          FQ223321.1 - 3, 4 219
          FQ223373.1 - 3, 4 237
          FQ223410.1 - 3, 4 335
          FQ223604.1 - 558
          FQ223661.1 - 3, 4 238
          FQ224433.1 - 3, 4 240
          FQ224506.1 - 3, 4 237
          FQ224779.1 - 3, 4 238
          FQ228655.1 - 3, 4 237
          FQ228991.1 - 3, 4 237
          FQ229525.1 - 3, 4 236
          K02932.1 - 2, 3 168
          Gene Symbol: LOC100362027
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002727709.3 XP_002727755.1 1 245
          Gene Symbol: LOC103690975
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008758743.1 XP_008756965.1 2 282
          Gene Symbol: RGD1562394
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_344225.3 XP_344226.1 1 179
          Gene Symbol: Rpl30l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_344178.3 XP_344179.2 1 168

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000989, BQ055546, L05095, BC095426, X79238, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009083, NM_001163485, BC100608, BC086891, BC092137, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0