Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HES5 SIRNA Assays ›
Gene Symbol:
Hes5
Gene Name:
hes family bHLH transcription factor 5
Gene Aliases:
-
Chromosome Location:
Chr.5: 172364421 - 172365188 on Build Rnor_6.0
UniGene ID:
Rn.22422
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
189604
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hes5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024383.1
NP_077359.1
2
260
GenBank mRNA
D12516.1
-
2
260
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001010926, BC087840, DQ272660.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cell adhesion
Notch signaling pathway
smoothened signaling pathway
brain development
positive regulation of cell proliferation
oligodendrocyte development
telencephalon development
glial cell fate commitment
forebrain radial glial cell differentiation
neural tube development
central nervous system myelination
neuron differentiation
positive regulation of BMP signaling pathway
regulation of myelination
auditory receptor cell differentiation
positive regulation of tyrosine phosphorylation of Stat3 protein
auditory receptor cell fate determination
camera-type eye development
regulation of cell differentiation
negative regulation of neuron differentiation
positive regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
cell maturation
positive regulation of smooth muscle cell proliferation
astrocyte differentiation
negative regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
regulation of neurogenesis
cartilage development
comma-shaped body morphogenesis
S-shaped body morphogenesis
specification of loop of Henle identity
metanephric nephron tubule morphogenesis
neuronal stem cell population maintenance
negative regulation of stem cell differentiation
negative regulation of pro-B cell differentiation
negative regulation of forebrain neuron differentiation
negative regulation of inner ear receptor cell differentiation
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
double-stranded DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0