Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDKN2A SIRNA Assays ›
          Gene Symbol:
          Cdkn2a
          Gene Name:
          cyclin-dependent kinase inhibitor 2A
          Gene Aliases:
          Arf, INK4A, MTS1, p16, p16Cdkn2a, p19ARF
          Chromosome Location:
          Chr.5: 107823323 - 107832405 on Build Rnor_6.0
          UniGene ID:
          Rn.48717
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 189946

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cdkn2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031550.1 NP_113738.1 3 581
          GenBank mRNA AF474974.1 - 622
          AF474975.1 - 622
          AF474976.1 - 3 557
          AF474977.1 - 3 548
          AY679727.1 - 622
          AY679728.1 - 3 557
          L81167.1 - 3 581

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          G1/S transition of mitotic cell cycle
          protein polyubiquitination
          negative regulation of cell-matrix adhesion
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          rRNA processing
          negative regulation of protein kinase activity
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell cycle arrest
          Ras protein signal transduction
          aging
          cell aging
          negative regulation of cell proliferation
          epidermis development
          apoptotic mitochondrial changes
          rRNA transcription
          response to organic substance
          response to organonitrogen compound
          regulation of G2/M transition of mitotic cell cycle
          regulation of gene expression
          response to organic cyclic compound
          cerebellum development
          negative regulation of cell growth
          negative regulation of B cell proliferation
          regulation of protein stability
          protein destabilization
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of protein binding
          negative regulation of immature T cell proliferation in thymus
          positive regulation of protein sumoylation
          negative regulation of mammary gland epithelial cell proliferation
          positive regulation of smooth muscle cell apoptotic process
          somatic stem cell population maintenance
          senescence-associated heterochromatin focus assembly
          negative regulation of phosphorylation
          response to drug
          glucose homeostasis
          positive regulation of apoptotic process
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of nucleocytoplasmic transport
          regulation of protein export from nucleus
          somatic stem cell division
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of ubiquitin-protein transferase activity
          positive regulation of apoptotic process involved in mammary gland involution
          cellular response to hydrogen peroxide
          protein K63-linked ubiquitination
          positive regulation of cell cycle arrest
          cellular senescence
          replicative senescence
          positive regulation of protein localization to nucleus
          positive regulation of signal transduction by p53 class mediator
          regulation of apoptotic DNA fragmentation
          negative regulation of proteolysis involved in cellular protein catabolic process
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of macrophage apoptotic process
          positive regulation of cellular senescence
          p53 binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          transcription factor binding
          protein kinase binding
          poly(A) RNA binding
          NF-kappaB binding
          ubiquitin-protein transferase inhibitor activity
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0