Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK5R1 SIRNA Assays ›
Gene Symbol:
Cdk5r1
Gene Name:
cyclin-dependent kinase 5, regulatory subunit 1 (p35)
Gene Aliases:
Cdk5r, p35
Chromosome Location:
Chr.10: 67862054 - 67863255 on Build Rnor_6.0
UniGene ID:
Rn.11213
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
190519
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cdk5r1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053891.1
NP_446343.1
1
880
GenBank mRNA
U50707.1
-
1
880
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009871, AK142868, AF121847, BC050768, AK169667, and others.
Panther Classification:
Molecular Function -
kinase activator
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
protein phosphorylation
neuron cell-cell adhesion
G-protein coupled acetylcholine receptor signaling pathway
axon guidance
axonal fasciculation
brain development
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cerebellum development
superior olivary nucleus maturation
hippocampus development
cerebral cortex radially oriented cell migration
layer formation in cerebral cortex
neuron differentiation
negative regulation of axon extension
neuron projection development
regulation of actin cytoskeleton organization
serine phosphorylation of STAT3 protein
ionotropic glutamate receptor signaling pathway
positive regulation of neuron apoptotic process
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
ephrin receptor signaling pathway
rhythmic process
axon extension
regulation of dendritic spine morphogenesis
regulation of microtubule cytoskeleton organization
positive regulation of cell cycle arrest
positive regulation of protein serine/threonine kinase activity
positive regulation of protein targeting to membrane
cyclin-dependent protein serine/threonine kinase activity
calcium ion binding
protein binding
kinase activity
cyclin-dependent protein kinase 5 activator activity
protein kinase binding
protein serine/threonine kinase activator activity
cadherin binding
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0