Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR3 SIRNA Assays ›
          Gene Symbol:
          Tlr3
          Gene Name:
          toll-like receptor 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 50016468 - 50031011 on Build Rnor_6.0
          UniGene ID:
          Rn.15273
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 191707

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tlr3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198791.1 NP_942086.1 4 709
          XM_008771266.1 XP_008769488.1 5 958
          XM_008771267.1 XP_008769489.1 5 878
          GenBank mRNA AB116229.1 - 4 709

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          toll-like receptor signaling pathway
          microglial cell activation involved in immune response
          MyD88-independent toll-like receptor signaling pathway
          defense response
          I-kappaB phosphorylation
          male gonad development
          response to virus
          positive regulation of type I interferon production
          positive regulation of chemokine production
          positive regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor signaling pathway
          toll-like receptor 3 signaling pathway
          positive regulation of type III interferon production
          cellular response to interferon-beta
          cellular response to drug
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          response to dsRNA
          positive regulation of interferon-gamma biosynthetic process
          positive regulation of chemokine biosynthetic process
          positive regulation of interferon-alpha biosynthetic process
          positive regulation of interferon-beta biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          cellular response to mechanical stimulus
          cellular response to interferon-gamma
          cellular response to dsRNA
          cellular response to exogenous dsRNA
          extrinsic apoptotic signaling pathway
          necroptotic signaling pathway
          double-stranded RNA binding
          receptor activity
          transmembrane signaling receptor activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline