Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLT1 SIRNA Assays ›
          Gene Symbol:
          FLT1
          Gene Name:
          fms related tyrosine kinase 1
          Gene Aliases:
          FLT, FLT-1, VEGFR-1, VEGFR1
          Chromosome Location:
          Chr.13: 28300346 - 28495128 on Build GRCh38
          UniGene ID:
          Hs.594454
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 192

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FLT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159920.1 NP_001153392.1 1, 2 357
          NM_001160030.1 NP_001153502.1 1, 2 357
          NM_001160031.1 NP_001153503.1 1, 2 357
          NM_002019.4 NP_002010.2 1, 2 357
          XM_011535014.1 XP_011533316.1 1, 2 357
          XM_017020485.1 XP_016875974.1 1, 2 357
          GenBank mRNA AF063657.2 - 1, 2 72
          AK292936.1 - 1, 2 357
          AK300392.1 - 1, 2 357
          AK309901.1 - 1, 2 357
          BC039007.1 - 1, 2 308 5722772
          DA851664.1 - 1, 2 357
          EU360600.1 - 1, 2 357
          EU368830.1 - 1, 2 72
          EU826561.1 - 1, 2 72
          EU826562.1 - 1, 2 72
          U01134.1 - 1, 2 321

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          angiogenesis
          response to hypoxia
          monocyte chemotaxis
          regulation of smooth muscle contraction
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cell proliferation
          positive regulation of phospholipase C activity
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          cell differentiation
          positive regulation of cell migration
          intracellular receptor signaling pathway
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          cellular response to vascular endothelial growth factor stimulus
          vascular endothelial growth factor receptor-1 signaling pathway
          vascular endothelial growth factor signaling pathway
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of angiogenesis
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          blood vessel morphogenesis
          embryonic morphogenesis
          positive regulation of smooth muscle cell proliferation
          transmembrane receptor protein tyrosine kinase activity
          vascular endothelial growth factor-activated receptor activity
          protein binding
          ATP binding
          growth factor binding
          VEGF-A-activated receptor activity
          VEGF-B-activated receptor activity
          placental growth factor-activated receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0